Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptcd2
Entrez Gene ID
310025
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
-
Chromosome Location
Chr.2: 29570581 - 29598285 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107648.1 NP_001101118.1 1 - 2 132 156
XM_006231847.2 XP_006231909.1 1 - 2 164 156

View Full Product Details