The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008760379.1 | XP_008758601.1 | 2 - 3 | 2356 | 132 |
XM_008760380.1 | XP_008758602.1 | 2 - 3 | 2356 | 132 | |
XM_008760381.1 | XP_008758603.1 | 2 - 3 | 1848 | 132 | |
XM_008760382.1 | XP_008758604.1 | 1 - 2 | 1067 | 132 | |
XM_008760383.1 | XP_008758605.1 | 2 - 3 | 1202 | 132 | |
XM_008760384.1 | XP_008758606.1 | 2 - 3 | 2356 | 132 |