Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a18
Entrez Gene ID
100359485
Gene Name
membrane-spanning 4-domains, subfamily A, member 18
Gene Aliases
-
Chromosome Location
Chr.1: 227050349 - 227071483 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760254.1 XP_008758476.1 6 - 7 1283 77

View Full Product Details