Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dbp
Entrez Gene ID
24309
Gene Name
D site of albumin promoter (albumin D-box) binding protein
Gene Aliases
-
Chromosome Location
Chr.1: 101687896 - 101692845 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012543.3 NP_036675.1 2 - 3 948 116
GenBank mRNA BC087668.1 - 2 - 3 948 116
FQ213620.1 - 2 - 3 462 116
J03179.1 - 2 - 3 914 116

View Full Product Details