Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klk4
Entrez Gene ID
408210
Gene Name
kallikrein-related peptidase 4
Gene Aliases
Emsp1
Chromosome Location
Chr.1: 99828944 - 99834642 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004101.1 NP_001004101.1 4 - 5 624 70

View Full Product Details