Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap14
Entrez Gene ID
501768
Gene Name
keratin associated protein 14
Gene Aliases
RGD1564302
Chromosome Location
Chr.11: 28776761 - 28777603 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109326.1 NP_001102796.1 1 - 1 633 112

View Full Product Details