Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnn
Entrez Gene ID
368070
Gene Name
pinin, desmosome associated protein
Gene Aliases
-
Chromosome Location
Chr.6: 80159364 - 80167677 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109023.1 NP_001102493.1 2 - 3 199 72

View Full Product Details