The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109023.1 | NP_001102493.1 | 7 - 8 | 675 | 133 |
GenBank mRNA | FQ234501.1 | - | 7 - 8 | 684 | 133 |