Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sult6b1
Entrez Gene ID
503103
Gene Name
sulfotransferase family, cytosolic, 6B, member 1
Gene Aliases
-
Chromosome Location
Chr.6: 1468083 - 1493670 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192017.1 NP_001178946.1 4 - 5 533 153

View Full Product Details