Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abhd1
Entrez Gene ID
313917
Gene Name
abhydrolase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 26787807 - 26792795 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008520.1 NP_001008520.1 1 - 2 182 71
GenBank mRNA BC086351.1 - 1 - 2 182 71

View Full Product Details