Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hadhb
Entrez Gene ID
171155
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit
Gene Aliases
TP-beta
Chromosome Location
Chr.6: 27555411 - 27589693 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006239786.1 XP_006239848.1 12 - 13 1538 132
XM_006239788.1 XP_006239850.1 12 - 13 1194 132
XM_006239789.2 XP_006239851.1 12 - 13 1469 132
GenBank mRNA BC060545.1 - 12 - 13 1121 132
D16479.1 - 11 - 12 1085 132
FM105991.1 - 3 - 4 265 132

View Full Product Details