The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017233.1 | NP_058929.1 | 11 - 12 | 864 | 57 |
GenBank mRNA | AF082834.1 | - | 11 - 12 | 864 | 57 |
BC081819.1 | - | 11 - 12 | 862 | 57 | |
FQ209393.1 | - | 10 - 11 | 1020 | 57 | |
FQ209489.1 | - | 10 - 11 | 825 | 57 | |
FQ210905.1 | - | 11 - 12 | 872 | 57 | |
FQ218378.1 | - | 11 - 12 | 873 | 57 | |
FQ218745.1 | - | 11 - 12 | 872 | 57 | |
FQ219696.1 | - | 11 - 12 | 872 | 57 | |
M18405.1 | - | 10 - 11 | 820 | 57 |