Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mvk
Entrez Gene ID
81727
Gene Name
mevalonate kinase
Gene Aliases
Lrbp
Chromosome Location
Chr.12: 47904266 - 47920457 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031063.1 NP_112325.1 5 - 6 608 76
GenBank mRNA M29472.1 - 5 - 6 608 76

View Full Product Details