Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syna
Entrez Gene ID
288602
Gene Name
syncytin a
Gene Aliases
Gm52
Chromosome Location
Chr.12: 25161130 - 25168182 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014771.2 NP_001014771.1 1 - 2 73 120

View Full Product Details