Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc15a4
Entrez Gene ID
246280
Gene Name
solute carrier family 15 (oligopeptide transporter), member 4
Gene Aliases
-
Chromosome Location
Chr.12: 32753824 - 32773277 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144758.1 NP_653359.1 4 - 5 1110 120
GenBank mRNA AB000280.1 - 4 - 5 1110 120
BC087709.1 - 4 - 5 1109 120

View Full Product Details