Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srrm3
Entrez Gene ID
685890
Gene Name
serine/arginine repetitive matrix 3
Gene Aliases
RGD1307391
Chromosome Location
Chr.12: 23855400 - 23925741 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277394.1 NP_001264323.1 2 - 3 390 62

View Full Product Details