Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mios
Entrez Gene ID
362324
Gene Name
missing oocyte, meiosis regulator, homolog (Drosophila)
Gene Aliases
RGD1308432
Chromosome Location
Chr.4: 34238381 - 34264015 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173554.1 NP_001167025.1 2 - 3 273 102

View Full Product Details