Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35b4
Entrez Gene ID
296969
Gene Name
solute carrier family 35 (UDP-xylose/UDP-N-acetylglucosamine transporter), member B4
Gene Aliases
-
Chromosome Location
Chr.4: 61548117 - 61573016 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106590.1 NP_001100060.1 1 - 2 297 75

View Full Product Details