Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klrg2
Entrez Gene ID
689885
Gene Name
killer cell lectin-like receptor subfamily G, member 2
Gene Aliases
-
Chromosome Location
Chr.4: 66370325 - 66381334 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001072378.4 XP_001072378.2 4 - 5 1647 104

View Full Product Details