The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109223.1 | NP_001102693.1 | 2 - 3 | 623 | 76 |
XM_006236143.2 | XP_006236205.1 | 2 - 3 | 672 | 76 |