Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hyal5
Entrez Gene ID
500052
Gene Name
hyaluronoglucosaminidase 5
Gene Aliases
-
Chromosome Location
Chr.4: 52232580 - 52275954 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024321.1 NP_001019492.1 2 - 3 1263 145
XM_006236166.2 XP_006236228.1 3 - 4 1381 145
XM_008762806.1 XP_008761028.1 4 - 5 1609 145
XM_008762807.1 XP_008761029.1 3 - 4 1516 145
XM_008762808.1 XP_008761030.1 3 - 4 1431 145
GenBank mRNA BC079447.1 - 4 - 5 1586 145
BC091219.1 - 2 - 3 1263 145

View Full Product Details