The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108624.2 | NP_001102094.2 | 2 - 3 | 278 | 102 |
GenBank mRNA | BC166901.1 | - | 2 - 3 | 255 | 102 |
BG668360.1 | - | 1 - 2 | 138 | 102 |