Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asz1
Entrez Gene ID
170578
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
Gasz
Chromosome Location
Chr.4: 45358574 - 45414177 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130750.1 NP_570106.1 5 - 6 573 136
GenBank mRNA AF461260.1 - 5 - 6 573 136

View Full Product Details