Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Casp2
Entrez Gene ID
64314
Gene Name
caspase 2
Gene Aliases
-
Chromosome Location
Chr.4: 71652362 - 71670228 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022522.2 NP_071967.2 5 - 6 576 64
XM_006236394.2 XP_006236456.1 4 - 5 1257 64
XM_008762918.1 XP_008761140.1 4 - 5 1257 64
GenBank mRNA AF136231.1 - 5 - 6 574 64
AF136232.1 - 5 - 6 574 64
U77933.1 - 5 - 6 576 64

View Full Product Details