Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr91
Entrez Gene ID
312225
Gene Name
WD repeat domain 91
Gene Aliases
RGD1306697
Chromosome Location
Chr.4: 62398073 - 62438958 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127298.2 NP_001120770.2 1 - 2 141 72
GenBank mRNA BC166785.1 - 1 - 2 141 72

View Full Product Details