Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhdc10
Entrez Gene ID
312199
Gene Name
kelch domain containing 10
Gene Aliases
RGD1309792
Chromosome Location
Chr.4: 57715979 - 57773800 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017456.1 NP_001017456.1 3 - 4 607 73
XM_006236201.2 XP_006236263.1 4 - 5 763 73
GenBank mRNA BC085351.1 - 3 - 4 607 73

View Full Product Details