Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Napg
Entrez Gene ID
307382
Gene Name
N-ethylmaleimide-sensitive factor attachment protein, gamma
Gene Aliases
-
Chromosome Location
Chr.18: 58325319 - 58344071 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107384.1 NP_001100854.1 4 - 5 374 147
XM_006254829.2 XP_006254891.1 5 - 6 366 142
GenBank mRNA AY724541.1 - 5 - 6 271 142

View Full Product Details