Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sirt3
Entrez Gene ID
293615
Gene Name
sirtuin 3
Gene Aliases
-
Chromosome Location
Chr.1: 213613502 - 213636194 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FQ218084.1 - 1 - 2 62 84

View Full Product Details