Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cfl1
Entrez Gene ID
29271
Gene Name
cofilin 1, non-muscle
Gene Aliases
-
Chromosome Location
Chr.1: 220869805 - 220873337 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017147.2 NP_058843.1 1 - 2 87 118
GenBank mRNA BC059143.1 - 1 - 2 87 118
BC086533.1 - 1 - 2 72 118
FQ212794.1 - 1 - 2 86 118
FQ212910.1 - 1 - 2 82 118
FQ213701.1 - 1 - 2 82 118
FQ213897.1 - 1 - 2 111 118
FQ214120.1 - 1 - 2 83 118
FQ214438.1 - 1 - 2 82 118
FQ220384.1 - 1 - 2 98 118
FQ220571.1 - 1 - 2 84 118
FQ226405.1 - 1 - 2 82 118
FQ227487.1 - 1 - 2 84 118
FQ229766.1 - 1 - 2 86 118
FQ229833.1 - 1 - 2 82 118
FQ229875.1 - 1 - 2 86 118
FQ230091.1 - 1 - 2 83 118
FQ231379.1 - 1 - 2 82 118
FQ233750.1 - 1 - 2 82 118
FQ234162.1 - 1 - 2 86 118
FQ234303.1 - 1 - 2 82 118

View Full Product Details