Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptdss2
Entrez Gene ID
293620
Gene Name
phosphatidylserine synthase 2
Gene Aliases
PSS-2
Chromosome Location
Chr.1: 214122989 - 214148534 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106316.1 NP_001099786.1 11 - 12 1358 58
XM_006230532.2 XP_006230594.1 11 - 12 2174 58
GenBank mRNA BC166496.1 - - 1303 58

View Full Product Details