The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133619.2 | NP_598303.1 | 3 - 4 | 500 | 123 |
XM_008760069.1 | XP_008758291.1 | 3 - 4 | 2536 | 123 | |
XM_008760070.1 | XP_008758292.1 | 3 - 4 | 2377 | 123 | |
GenBank mRNA | AF260741.1 | - | 3 - 4 | 500 | 123 |