Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Athl1
Entrez Gene ID
309103
Gene Name
ATH1, acid trehalase-like 1 (yeast)
Gene Aliases
RGD1306015
Chromosome Location
Chr.1: 213734206 - 213743975 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230587.2 XP_006230649.1 3 - 4 975 116

View Full Product Details