The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130408.2 | NP_569092.2 | 5 - 6 | 1036 | 121 |
GenBank mRNA | AF439720.2 | - | 5 - 6 | 1036 | 121 |
DQ266888.1 | - | 5 - 6 | 1106 | 121 | |
DQ317305.1 | - | 5 - 6 | 1046 | 121 |