The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212514.2 | NP_997679.1 | 3 - 4 | 363 | 71 |
XM_006241974.1 | XP_006242036.1 | 4 - 5 | 452 | 71 | |
GenBank mRNA | AB162854.1 | - | 3 - 4 | 350 | 71 |
BC098639.1 | - | 3 - 4 | 363 | 71 | |
FQ213035.1 | - | 1 - 2 | 31 | 71 | |
FQ225246.1 | - | 1 - 2 | 31 | 71 | |
FQ227798.1 | - | 1 - 2 | 31 | 71 | |
FQ230071.1 | - | 1 - 2 | 31 | 71 | |
FQ234651.1 | - | 1 - 2 | 31 | 71 |