Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnj4
Entrez Gene ID
116649
Gene Name
potassium channel, inwardly rectifying subfamily J, member 4
Gene Aliases
Hirk2
Chromosome Location
Chr.7: 120717378 - 120744602 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053870.2 NP_446322.2 1 - 2 551 95
GenBank mRNA X87635.1 - 1 - 2 551 95

View Full Product Details