Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dscc1
Entrez Gene ID
299933
Gene Name
DNA replication and sister chromatid cohesion 1
Gene Aliases
RGD1561749
Chromosome Location
Chr.7: 94759076 - 94774784 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_216934.8 XP_216934.3 2 - 3 627 157

View Full Product Details