Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ly6c
Entrez Gene ID
56778
Gene Name
Ly6-C antigen
Gene Aliases
-
Chromosome Location
Chr.7|NW_007906224: 20092 - 23957 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257716.2 XP_006257778.1 1 - 2 71 74
GenBank mRNA FQ230335.1 - 1 - 2 25 74
FQ232627.1 - 1 - 2 25 74
FQ235068.1 - 1 - 2 25 74

Gene Symbol
LOC100911104
Entrez Gene ID
100911104
Gene Name
lymphocyte antigen 6B-like
Gene Aliases
-
Chromosome Location
Chr.14: 22868518 - 22872383 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751343.2 XP_003751391.1 1 - 2 71 74

View Full Product Details