The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128494.1 | NP_001121966.1 | 3 - 4 | 710 | 74 |
GenBank mRNA | BC166737.1 | - | 3 - 4 | 710 | 74 |