Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prdm4
Entrez Gene ID
170820
Gene Name
PR domain containing 4
Gene Aliases
SC-1
Chromosome Location
Chr.7: 23959929 - 23986535 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133312.1 NP_579846.1 3 - 4 1118 102
XM_006241160.2 XP_006241222.1 5 - 6 1580 102
XM_006241161.1 XP_006241223.1 5 - 6 1579 102
XM_008765213.1 XP_008763435.1 5 - 6 1585 102
GenBank mRNA AF176023.1 - 3 - 4 1118 102

View Full Product Details