Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc59
Entrez Gene ID
314799
Gene Name
coiled-coil domain containing 59
Gene Aliases
RGD1311912
Chromosome Location
Chr.7: 47928138 - 47934690 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108090.1 NP_001101560.1 3 - 4 978 152

View Full Product Details