Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nap1l1
Entrez Gene ID
89825
Gene Name
nucleosome assembly protein 1-like 1
Gene Aliases
-
Chromosome Location
Chr.7: 54213305 - 54237880 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053561.2 NP_446013.2 14 - 15 1306 86
XM_006241342.1 XP_006241404.1 14 - 15 1320 86
XM_006241344.2 XP_006241406.1 12 - 13 1057 86
GenBank mRNA AF062594.1 - 14 - 15 1288 86
CA511505.1 - 3 - 4 137 86
CF110129.1 - 9 - 10 741 86
FQ230040.1 - 5 - 6 353 86

View Full Product Details