Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nap1l1
Entrez Gene ID
89825
Gene Name
nucleosome assembly protein 1-like 1
Gene Aliases
-
Chromosome Location
Chr.7: 54213305 - 54237880 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053561.2 NP_446013.2 5 - 6 512 129
XM_006241342.1 XP_006241404.1 5 - 6 526 129
XM_006241343.2 XP_006241405.1 5 - 6 526 129
XM_006241344.2 XP_006241406.1 3 - 4 263 129
GenBank mRNA AF062594.1 - 5 - 6 497 129

View Full Product Details