Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nup37
Entrez Gene ID
299706
Gene Name
nucleoporin 37
Gene Aliases
-
Chromosome Location
Chr.7: 28715298 - 28750978 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106775.1 NP_001100245.1 2 - 3 395 101
XM_006241197.1 XP_006241259.1 2 - 3 300 101
XM_006241198.2 XP_006241260.1 2 - 3 295 101
GenBank mRNA BC091253.1 - 1 - 2 157 101
BC161937.1 - 2 - 3 267 101
FQ220712.1 - 2 - 3 278 101

View Full Product Details