Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krr1
Entrez Gene ID
314830
Gene Name
KRR1, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
Hrb2
Chromosome Location
Chr.7: 54756043 - 54767477 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108094.2 NP_001101564.1 1 - 2 225 72
GenBank mRNA BC128768.1 - 1 - 2 225 72

View Full Product Details