Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab3ip
Entrez Gene ID
29885
Gene Name
RAB3A interacting protein
Gene Aliases
RABIN3
Chromosome Location
Chr.7: 59927486 - 59971516 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017313.3 NP_059009.2 8 - 9 1255 139
XM_006241357.2 XP_006241419.1 9 - 10 1424 139
XM_006241358.2 XP_006241420.1 9 - 10 1429 139
XM_006241359.2 XP_006241421.1 9 - 10 2433 139
XM_006241360.2 XP_006241422.1 9 - 10 1419 139
XM_008765382.1 XP_008763604.1 7 - 8 1385 139
XM_008765383.1 XP_008763605.1 8 - 9 864 139
GenBank mRNA BC127500.1 - 8 - 9 1258 139
U19181.1 - 8 - 9 1248 139

View Full Product Details