Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ndufa4l2
Entrez Gene ID
100362331
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2
Gene Aliases
-
Chromosome Location
Chr.7: 70821904 - 70825017 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271272.1 NP_001258201.1 2 - 3 147 79
XM_008765373.1 XP_008763595.1 2 - 3 200 79
GenBank mRNA FM120013.1 - 2 - 3 150 79

View Full Product Details