Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aco2
Entrez Gene ID
79250
Gene Name
aconitase 2, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.7: 123102493 - 123145608 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024398.2 NP_077374.2 14 - 15 1770 72
GenBank mRNA BC061999.1 - 14 - 15 1770 72

View Full Product Details