The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277194.1 | NP_001264123.1 | 6 - 7 | 909 | 152 |
GenBank mRNA | FM122421.1 | - | 3 - 4 | 434 | 152 |
FQ212206.1 | - | 3 - 4 | 351 | 152 | |
FQ214725.1 | - | 6 - 7 | 890 | 152 | |
FQ216469.1 | - | 6 - 7 | 893 | 152 | |
FQ220496.1 | - | 6 - 7 | 886 | 152 | |
FQ220832.1 | - | 6 - 7 | 892 | 152 | |
FQ222191.1 | - | 6 - 7 | 891 | 152 | |
FQ222396.1 | - | 6 - 7 | 891 | 152 | |
FQ224965.1 | - | 6 - 7 | 888 | 152 |