Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc38
Entrez Gene ID
300125
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
RGD1306041
Chromosome Location
Chr.7: 126711659 - 126735013 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130499.1 NP_001123971.1 6 - 7 690 79
GenBank mRNA FQ210197.1 - 6 - 7 659 79
FQ218771.1 - 6 - 7 700 79

View Full Product Details