Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc45a4
Entrez Gene ID
315054
Gene Name
solute carrier family 45, member 4
Gene Aliases
RGD1309971
Chromosome Location
Chr.7: 114820718 - 114888982 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006241741.2 XP_006241803.1 1 - 2 113 75

View Full Product Details